Assay Details
Target Gene Details
Entrez Gene ID: | 93129 |
Gene Name: | ORAI calcium release-activated calcium modulator 3 |
Gene Aliases: |
TMEM142C |
Location: |
Chr.16:30949084-30954938 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORAI3 | NM_152288.2 | 2 | 620 | NP_689501.1 |
| AK298276.1 | 2 | 582 | ||
| BC006126.1 | 2 | 563 | AAH06126.2 | |
| BC015555.1 | 2 | 609 | AAH15555.1 | |
| BC022786.1 | 2 | 563 | AAH22786.1 | |
| BI561433.1 | 2 | 641 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482192 | Chr.16:30949084 - 30954937 on Build GRCh38 | Gain |
|
| dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss |
|
| nsv571820 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
| nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion |
|
| nsv833193 | Chr.16:30949475 - 31122283 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map