Assay Details
Target Gene Details
Entrez Gene ID: | 286207 |
Gene Name: | cilia and flagella associated protein 157 |
Gene Aliases: |
C9orf117 |
Location: |
Chr.9:127706261-127716002 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP157 | XM_011518559.2 | 8 | 2548 | XP_011516861.2 |
| AK126437.1 | 4 | 1853 | ||
| AL833241.1 | 2 | 2432 | ||
| BC133027.1 | 2 | 1459 |
Target Gene Details
Entrez Gene ID: | 138428 |
Gene Name: | peptidyl-tRNA hydrolase 1 homolog |
Gene Aliases: |
C9orf115, PTH1 |
Location: |
Chr.9:127695024-127716714 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTRH1 | XM_006716955.3 | 1 | 989 | XP_006717018.1 |
| XM_017014276.1 | 1 | 989 | XP_016869765.1 | |
| XM_017014277.1 | 1 | 989 | XP_016869766.1 | |
| XM_017014278.1 | 1 | 989 | XP_016869767.1 | |
| AK090922.1 | 2 | 1371 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615369 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map