Assay Details
Target Gene Details
Entrez Gene ID: | 100128494 |
Gene Name: | uncharacterized LOC100128494 |
Gene Aliases: |
- |
Location: |
Chr.11:72014291-72020910 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100128494 | NR_104178.1 | |||
| AK126629.1 |
Target Gene Details
Entrez Gene ID: | 4926 |
Gene Name: | nuclear mitotic apparatus protein 1 |
Gene Aliases: |
NMP-22, NUMA |
Location: |
Chr.11:72002864-72080693 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUMA1 | NM_001286561.1 | 11 | 1090 | NP_001273490.1 |
| NM_006185.3 | 10 | 992 | NP_006176.2 | |
| NR_104476.1 | 10 | 911 | ||
| XM_006718564.1 | 9 | 807 | XP_006718627.1 | |
| XM_011545054.1 | 9 | 807 | XP_011543356.1 | |
| XM_011545055.1 | 10 | 985 | XP_011543357.1 | |
| XM_011545056.2 | 11 | 979 | XP_011543358.1 | |
| XM_011545057.2 | 11 | 969 | XP_011543359.1 | |
| XM_011545058.1 | 9 | 1006 | XP_011543360.1 | |
| XM_011545059.2 | 11 | 971 | XP_011543361.1 | |
| XM_011545060.2 | 11 | 965 | XP_011543362.1 | |
| XM_011545061.2 | 9 | 825 | XP_011543363.1 | |
| XM_011545062.2 | 9 | 838 | XP_011543364.1 | |
| XM_011545063.2 | 9 | 805 | XP_011543365.1 | |
| XM_011545064.1 | 9 | 792 | XP_011543366.1 | |
| XM_011545065.1 | 9 | 807 | XP_011543367.1 | |
| XM_011545066.1 | 9 | 807 | XP_011543368.1 | |
| XM_017017830.1 | 10 | 847 | XP_016873319.1 | |
| XM_017017831.1 | 9 | 843 | XP_016873320.1 | |
| AB210007.1 | 10 | 793 | ||
| AK290509.1 | 9 | 807 | ||
| AK307577.1 | 10 | 810 | ||
| AK309041.1 | 9 | 814 | ||
| BC004165.2 | 10 | 826 | AAH04165.1 | |
| BC027493.1 | 10 | 793 | ||
| BC043499.1 | 10 | 793 | AAH43499.1 | |
| BC103765.1 | 10 | 911 | AAI03766.1 | |
| Z11583.1 | 10 | 810 | CAA77669.1 | |
| Z11584.1 | 11 | 912 | CAA77670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map