Assay Details
Target Gene Details
Entrez Gene ID: | 80196 |
Gene Name: | ring finger protein 34 |
Gene Aliases: |
CARP-1, CARP1, RFI, RIF, RIFF, hRFI |
Location: |
Chr.12:121400083-121424353 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF34 | NM_001256858.1 | NP_001243787.1 | ||
| NM_025126.3 | NP_079402.2 | |||
| NM_194271.2 | NP_919247.1 | |||
| AF306709.1 | AAK51328.1 | |||
| AK025439.1 | BAB15132.1 | |||
| AK225285.1 | ||||
| AK304366.1 | ||||
| AL557811.3 | ||||
| BC007826.2 | AAH07826.1 | |||
| BI559962.1 | ||||
| DA887995.1 | ||||
| DB242808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv522e212 | Chr.12:121387834 - 121410687 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map