Assay Details
Target Gene Details
Entrez Gene ID: | 83468 |
Gene Name: | glycosyltransferase 8 domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:103988983-104064177 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLT8D2 | NM_001316967.1 | 9 | 1330 | NP_001303896.1 |
| NM_031302.4 | 10 | 1465 | NP_112592.1 | |
| XM_011538793.2 | 10 | 1700 | XP_011537095.1 | |
| XM_017019999.1 | 11 | 1781 | XP_016875488.1 | |
| AJ271711.1 | 7 | 679 | CAC19667.1 | |
| AK027310.1 | 8 | 779 | BAB55033.1 | |
| AK056056.1 | 9 | 1336 | BAB71085.1 | |
| AY358763.1 | 9 | 869 | AAQ89123.1 | |
| BC022343.1 | 9 | 1086 | AAH22343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560039 | Chr.12:103783890 - 104061562 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map