Assay Details
Target Gene Details
Entrez Gene ID: | 23017 |
Gene Name: | Fas apoptotic inhibitory molecule 2 |
Gene Aliases: |
LFG, LFG2, NGP35, NMP35, TMBIM2 |
Location: |
Chr.12:49866896-49904275 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAIM2 | NM_012306.3 | 12 | 4551 | NP_036438.2 |
| XM_005268730.3 | 11 | 4294 | XP_005268787.1 | |
| XM_017019040.1 | 13 | 4658 | XP_016874529.1 | |
| AB023167.1 | 12 | 4448 | ||
| BX647219.1 | 2 | 3137 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832407 | Chr.12:49849485 - 50018453 on Build GRCh38 | Loss |
|
| nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain |
|
| nsv952178 | Chr.12:49866318 - 49896717 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map