Assay Details
Target Gene Details
Entrez Gene ID: | 27336 |
Gene Name: | HIV-1 Tat specific factor 1 |
Gene Aliases: |
TAT-SF1, TATSF1, dJ196E23.2 |
Location: |
Chr.X:136496932-136512346 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTATSF1 | NM_001163280.1 | 10 | 2098 | NP_001156752.1 |
| NM_014500.4 | 9 | 1849 | NP_055315.2 | |
| XM_005262404.3 | 10 | 2193 | XP_005262461.1 | |
| AB209303.1 | 10 | 2098 | BAD92540.1 | |
| AK225870.1 | 9 | 1796 | ||
| AK299403.1 | 9 | 1801 | ||
| BC009896.2 | 9 | 1849 | AAH09896.1 | |
| BT006886.1 | 9 | 1676 | AAP35532.1 | |
| U76992.1 | 9 | 1733 | AAB18823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577409 | Chr.X:136508165 - 136512467 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map