Assay Details
Target Gene Details
Entrez Gene ID: | 10397 |
Gene Name: | N-myc downstream regulated 1 |
Gene Aliases: |
CAP43, CMT4D, DRG-1, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5 |
Location: |
Chr.8:133237171-133297587 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG1 | NM_001135242.1 | 16 | 2789 | NP_001128714.1 |
| NM_001258432.1 | 14 | 2275 | NP_001245361.1 | |
| NM_001258433.1 | 15 | 2286 | NP_001245362.1 | |
| NM_006096.3 | 16 | 2392 | NP_006087.2 | |
| XM_011516791.1 | 16 | 2726 | XP_011515093.1 | |
| XM_011516792.1 | 9 | 2625 | XP_011515094.1 | |
| AF004162.1 | 15 | 2231 | AAC13419.1 | |
| AK091147.1 | 15 | 2235 | ||
| AK091269.1 | ||||
| AK095649.1 | 14 | 2128 | BAC04597.1 | |
| AK096562.1 | 16 | 2227 | ||
| AK124709.1 | 8 | 3299 | ||
| AK126924.1 | 16 | 3071 | ||
| AK129867.1 | 7 | 1666 | ||
| BC003175.1 | 16 | 2320 | AAH03175.1 | |
| D87953.1 | 16 | 2325 | BAA13505.1 | |
| X92845.1 | 16 | 2313 | CAA63430.1 |
More Information
Set Membership: |
|

Genomic Map