Assay Details
Target Gene Details
Entrez Gene ID: | 9057 |
Gene Name: | solute carrier family 7 member 6 |
Gene Aliases: |
LAT-2, LAT3, y+LAT-2 |
Location: |
Chr.16:68263830-68301823 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A6 | NM_001076785.2 | 4 | 642 | NP_001070253.1 |
| NM_003983.5 | 3 | 555 | NP_003974.3 | |
| XM_011523433.1 | 3 | 657 | XP_011521735.1 | |
| XM_011523434.1 | 3 | 722 | XP_011521736.1 | |
| XM_011523438.1 | 2 | 1112 | XP_011521740.1 | |
| XM_011523439.1 | 3 | 1241 | XP_011521741.1 | |
| XM_017023851.1 | XP_016879340.1 | |||
| AK310866.1 | 3 | 521 | ||
| AK311610.1 | 1 | 187 | ||
| BC028216.1 | 3 | 511 | AAH28216.1 | |
| BC113100.1 | 3 | 542 | ||
| CR749291.1 | 3 | 541 | CAH18146.1 | |
| CR749475.1 | 3 | 524 | CAH18305.1 | |
| D87432.1 | 3 | 590 | ||
| DB107640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510691 | Chr.16:68266527 - 68325431 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map