Assay Details
Target Gene Details
Entrez Gene ID: | 282974 |
Gene Name: | serine/threonine kinase 32C |
Gene Aliases: |
PKE, YANK3 |
Location: |
Chr.10:132207482-132332559 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK32C | NM_001318878.1 | NP_001305807.1 | ||
| NM_001318879.1 | NP_001305808.1 | |||
| NM_173575.3 | NP_775846.2 | |||
| NR_134911.1 | ||||
| XM_011539688.1 | XP_011537990.1 | |||
| XM_011539690.2 | XP_011537992.1 | |||
| XM_011539693.2 | XP_011537995.1 | |||
| XM_011539694.1 | XP_011537996.1 | |||
| XM_011539695.1 | XP_011537997.1 | |||
| XM_011539696.1 | XP_011537998.1 | |||
| AK057849.1 | ||||
| AK098366.1 | 5 | 740 | ||
| AK302102.1 | ||||
| AK308956.1 | ||||
| AK310725.1 | 6 | 907 | ||
| AY098866.1 | AAM21719.1 | |||
| BC045760.1 | AAH45760.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054569 | Chr.10:132009350 - 132351508 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| esv2761642 | Chr.10:132048518 - 132365985 on Build GRCh38 | Gain |
|
| nsv552504 | Chr.10:132180474 - 132255847 on Build GRCh38 | Loss |
|
| nsv552502 | Chr.10:132139965 - 132247418 on Build GRCh38 | Loss |
|
| nsv951886 | Chr.10:132219797 - 132258596 on Build GRCh38 | Deletion |
|
| nsv832030 | Chr.10:132093129 - 132236507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map