Assay Details
Target Gene Details
Entrez Gene ID: | 9223 |
Gene Name: | membrane associated guanylate kinase, WW and PDZ domain containing 1 |
Gene Aliases: |
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3 |
Location: |
Chr.3:65353525-66038836 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 25 |
More Information
Set Membership: |
|

Genomic Map