Assay Details
Target Gene Details
Entrez Gene ID: | 6204 |
Gene Name: | ribosomal protein S10 |
Gene Aliases: |
DBA9, S10 |
Location: |
Chr.6:34417454-34426125 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS10 | NM_001014.4 | NP_001005.1 | ||
| NM_001203245.2 | NP_001190174.1 | |||
| NM_001204091.1 | NP_001191020.1 | |||
| AK311797.1 | ||||
| BC001032.2 | AAH01032.1 | |||
| BC001955.1 | AAH01955.1 | |||
| BC005012.1 | AAH05012.1 | |||
| BC070235.1 | AAH70235.1 | |||
| BC071946.1 | AAH71946.1 | |||
| BC073799.1 | AAH73799.1 | |||
| BE397113.1 | ||||
| BG532491.1 | ||||
| BG940847.1 | ||||
| BU956627.1 | ||||
| BY996439.1 | ||||
| CD385233.1 |
Target Gene Details
Entrez Gene ID: | 100529239 |
Gene Name: | RPS10-NUDT3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.6:34287194-34426125 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS10-NUDT3 | NM_001202470.2 | NP_001189399.1 | ||
| BU521288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv981133 | Chr.6:34417379 - 34425744 on Build GRCh38 | Duplication |
|
| esv2759418 | Chr.6:34414715 - 34746900 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map