Assay Details
Target Gene Details
Entrez Gene ID: | 5265 |
Gene Name: | serpin family A member 1 |
Gene Aliases: |
A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT |
Location: |
Chr.14:94376747-94390692 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINA1 | NM_001002235.2 | NP_001002235.1 | ||
| NM_001002236.2 | 2 | 317 | NP_001002236.1 | |
| NM_001127700.1 | NP_001121172.1 | |||
| NM_001127701.1 | 2 | 354 | NP_001121173.1 | |
| NM_001127702.1 | NP_001121174.1 | |||
| NM_001127703.1 | 2 | 317 | NP_001121175.1 | |
| NM_001127704.1 | 2 | 317 | NP_001121176.1 | |
| NM_001127705.1 | 2 | 317 | NP_001121177.1 | |
| NM_001127706.1 | NP_001121178.1 | |||
| NM_001127707.1 | NP_001121179.1 | |||
| XM_017021370.1 | XP_016876859.1 | |||
| AA317159.1 | ||||
| AK315637.1 | ||||
| BG483969.1 | ||||
| BP296710.1 | ||||
| BP300943.1 | ||||
| BP301274.1 | ||||
| BX161449.1 | CAD61914.1 | |||
| BX247968.1 | 2 | 244 | CAD62306.1 | |
| CD685665.1 | 2 | 202 | ||
| DA427262.1 | ||||
| DA428299.1 | 2 | 320 | ||
| DA433050.1 | ||||
| DA433058.1 | 2 | 260 | ||
| DA672472.1 | 2 | 297 | ||
| DA675981.1 | 2 | 259 | ||
| X05826.1 | 2 | 288 | CAA29267.1 | |
| X05827.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832863 | Chr.14:94348482 - 94501487 on Build GRCh38 | Gain |
|
| nsv509545 | Chr.14:94360156 - 94400412 on Build GRCh38 | Insertion |
|
| nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain |
|
| nsv1406 | Chr.14:94361166 - 94394512 on Build GRCh38 | Insertion |
|
| nsv832864 | Chr.14:94362101 - 94538150 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map