Assay Details
Target Gene Details
Entrez Gene ID: | 9970 |
Gene Name: | nuclear receptor subfamily 1 group I member 3 |
Gene Aliases: |
CAR, CAR1, MB67 |
Location: |
Chr.1:161229666-161238623 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Target Gene Details
Entrez Gene ID: | 84134 |
Gene Name: | translocase of outer mitochondrial membrane 40 like |
Gene Aliases: |
TOMM40B |
Location: |
Chr.1:161225939-161230746 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOMM40L | NM_001286373.1 | 9 | 1882 | NP_001273302.1 |
| NM_001286374.1 | 8 | 1789 | NP_001273303.1 | |
| NM_032174.5 | 10 | 1984 | NP_115550.2 | |
| XM_006711572.2 | 10 | 1859 | XP_006711635.1 | |
| XM_011510057.2 | 8 | 1552 | XP_011508359.1 | |
| XM_017002481.1 | 10 | 1822 | XP_016857970.1 | |
| XM_017002482.1 | 9 | 1750 | XP_016857971.1 | |
| AK022832.1 | 7 | 1600 | BAB14265.1 | |
| AK027704.1 | 10 | 1863 | BAB55309.1 | |
| BC014946.1 | 9 | 1837 | AAH14946.1 | |
| BX537951.1 | 10 | 1861 | CAD97918.1 |
More Information
Set Membership: |
|

Genomic Map