Assay Details
Target Gene Details
Entrez Gene ID: | 80835 |
Gene Name: | taste 1 receptor member 1 |
Gene Aliases: |
GM148, GPR70, T1R1, TR1 |
Location: |
Chr.1:6555278-6579757 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAS1R1 | NM_138697.3 | 3 | 722 | NP_619642.2 |
| NM_177540.2 | NP_803884.1 | |||
| XM_011542203.2 | 3 | 1088 | XP_011540505.1 | |
| XM_011542206.2 | 2 | 1274 | XP_011540508.1 | |
| XM_017002402.1 | 2 | 1274 | XP_016857891.1 | |
| XM_017002403.1 | XP_016857892.1 | |||
| AF387617.1 | 2 | 403 | AAL91358.1 | |
| AF387618.1 | 2 | 435 | AAL91359.1 | |
| AF387619.1 | AAL91360.1 | |||
| AF387620.1 | AAL91361.1 | |||
| AK292014.1 | ||||
| AK293499.1 | ||||
| AK315805.1 | 3 | 691 | ||
| BC136515.1 | 3 | 722 | ||
| BC136516.1 | 3 | 721 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831647 | Chr.1:6525809 - 6587530 on Build GRCh38 | Loss |
|
| nsv7880 | Chr.1:6417968 - 6611914 on Build GRCh38 | Gain+Loss |
|
| nsv545268 | Chr.1:6490316 - 6578004 on Build GRCh38 | Loss |
|
| nsv545269 | Chr.1:6546548 - 6630080 on Build GRCh38 | Loss |
|
| nsv831536 | Chr.1:6429326 - 6605343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map