Assay Details
Target Gene Details
Entrez Gene ID: | 344805 |
Gene Name: | transmembrane protease, serine 7 |
Gene Aliases: |
- |
Location: |
Chr.3:112039067-112081269 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS7 | NM_001042575.2 | NP_001036040.2 | ||
| NR_026734.1 | ||||
| XM_011512754.1 | XP_011511056.1 | |||
| XM_011512758.1 | XP_011511060.1 | |||
| AK131211.1 | BAD18401.1 | |||
| BC117322.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs150219803] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map