Assay Details
Target Gene Details
Entrez Gene ID: | 493 |
Gene Name: | ATPase plasma membrane Ca2+ transporting 4 |
Gene Aliases: |
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x |
Location: |
Chr.1:203626787-203744081 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP2B4 | NM_001001396.2 | 1 | 95 | NP_001001396.1 |
| NM_001684.4 | 1 | 95 | NP_001675.3 | |
| AL706212.1 | 1 | 82 | ||
| BX537444.1 | 1 | 322 | CAD97686.1 | |
| CR749393.1 | CAH18241.1 | |||
| CR936708.1 | 1 | 49 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762217 | Chr.1:203409626 - 203630064 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map