Assay Details
Target Gene Details
Entrez Gene ID: | 80142 |
Gene Name: | prostaglandin E synthase 2 |
Gene Aliases: |
C9orf15, GBF-1, GBF1, PGES2, mPGES-2 |
Location: |
Chr.9:128120693-128128462 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES2 | NM_001256335.1 | NP_001243264.1 | ||
| NM_025072.6 | NP_079348.1 | |||
| NM_198938.2 | NP_945176.1 | |||
| XM_005252239.2 | XP_005252296.1 | |||
| XM_011519050.1 | XP_011517352.1 | |||
| XM_011519051.2 | XP_011517353.1 | |||
| AK024100.1 | BAB14826.1 | |||
| AK057049.1 | ||||
| AK223520.1 | BAD97240.1 | |||
| AK301618.1 | ||||
| AL525982.3 | ||||
| BC009397.2 | AAH09397.2 | |||
| BC009456.1 | AAH09456.1 | |||
| BC011613.2 | AAH11613.1 | |||
| BG481657.1 | ||||
| BI826720.1 | ||||
| BM739176.1 | ||||
| BX493085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951769 | Chr.9:128101422 - 128129321 on Build GRCh38 | Deletion |
|
| nsv615383 | Chr.9:128091848 - 128128781 on Build GRCh38 | Loss |
|
| nsv818724 | Chr.9:128100434 - 128128781 on Build GRCh38 | Loss |
|
| nsv6723 | Chr.9:128106647 - 128141731 on Build GRCh38 | Deletion |
|
| nsv471324 | Chr.9:128039877 - 128130666 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map