Assay Details
Target Gene Details
Entrez Gene ID: | 51013 |
Gene Name: | exosome component 1 |
Gene Aliases: |
CGI-108, CSL4, Csl4p, SKI4, Ski4p, p13 |
Location: |
Chr.10:97435909-97446065 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC1 | NM_001318362.1 | NP_001305291.1 | ||
| NM_001318363.1 | NP_001305292.1 | |||
| NM_001318364.1 | NP_001305293.1 | |||
| NM_001318365.1 | NP_001305294.1 | |||
| NM_001318366.1 | NP_001305295.1 | |||
| NM_016046.4 | NP_057130.1 | |||
| NR_134586.1 | ||||
| NR_134587.1 | ||||
| XM_011539847.2 | XP_011538149.1 | |||
| XM_017016304.1 | XP_016871793.1 | |||
| AF151866.1 | AAD34103.1 | |||
| AK307229.1 | ||||
| AK313717.1 | ||||
| AW954858.1 | ||||
| BC022067.2 | AAH22067.1 | |||
| BI550688.1 | ||||
| BM692452.1 | ||||
| BU569997.1 | ||||
| DB137484.1 | ||||
| N34159.1 | ||||
| W39427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624313 | Chr.10:97431212 - 97441841 on Build GRCh38 | Gain |
|
| nsv552025 | Chr.10:97375027 - 97445034 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map