Assay Details
Target Gene Details
Entrez Gene ID: | 5502 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 1A |
Gene Aliases: |
I1, IPP1 |
Location: |
Chr.12:54579240-54588659 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R1A | NM_006741.3 | NP_006732.3 | ||
| XM_005268995.4 | XP_005269052.1 | |||
| XM_006719471.3 | XP_006719534.1 | |||
| AK123969.1 | BAC85737.1 | |||
| AY063766.1 | AAL48321.1 | |||
| BC022470.1 | AAH22470.1 | |||
| BP211831.1 | ||||
| CR457003.1 | CAG33284.1 | |||
| U48707.1 | AAB02402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832421 | Chr.12:54522913 - 54691618 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map