Assay Details
Target Gene Details
Entrez Gene ID: | 63895 |
Gene Name: | piezo type mechanosensitive ion channel component 2 |
Gene Aliases: |
C18orf30, C18orf58, DA3, DA5, FAM38B, FAM38B2, HsT748, HsT771, MWKS |
Location: |
Chr.18:10670237-11149561 on Build GRCh38 |
Assay Gene Location: | Within Exon 46 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIEZO2 | NM_022068.3 | 43 | 6943 | NP_071351.2 |
| XM_011525723.2 | 46 | 7873 | XP_011524025.1 | |
| XM_011525724.2 | 45 | 7816 | XP_011524026.1 | |
| XM_011525725.2 | 44 | 7783 | XP_011524027.1 | |
| XM_011525726.2 | 45 | 7690 | XP_011524028.1 | |
| XM_017025918.1 | 45 | 7834 | XP_016881407.1 | |
| AK027056.1 | 3 | 350 | BAB15641.1 | |
| AK302764.1 | 5 | 780 | ||
| JN790819.1 | 43 | 6777 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain |
|
| nsv576477 | Chr.18:10681714 - 10817568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map