Assay Details
Target Gene Details
Entrez Gene ID: | 11012 |
Gene Name: | kallikrein related peptidase 11 |
Gene Aliases: |
PRSS20, TLSP |
Location: |
Chr.19:51022231-51028034 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK11 | NM_001136032.2 | 6 | 879 | NP_001129504.1 |
| NM_001167605.1 | 6 | 966 | NP_001161077.1 | |
| NM_006853.2 | 6 | 891 | NP_006844.1 | |
| NM_144947.1 | 6 | 973 | NP_659196.1 | |
| XM_005258439.3 | 4 | 612 | XP_005258496.1 | |
| XM_011526369.1 | 6 | 1117 | XP_011524671.1 | |
| XM_011526370.2 | 6 | 991 | XP_011524672.1 | |
| XM_011526371.2 | 7 | 1111 | XP_011524673.1 | |
| XM_011526372.2 | 6 | 1024 | XP_011524674.1 | |
| XM_011526373.1 | 6 | 894 | XP_011524675.1 | |
| AB012917.1 | 6 | 886 | BAA33404.1 | |
| AB013730.1 | 6 | 885 | BAA88713.1 | |
| AB041036.1 | 6 | 973 | BAA96797.1 | |
| AB078780.1 | BAC54105.1 | |||
| AB259014.1 | 6 | 877 | ||
| AB261897.1 | ||||
| AY359014.1 | 6 | 870 | AAQ89373.1 | |
| BC022068.1 | 6 | 897 | AAH22068.1 |
More Information
Set Membership: |
|

Genomic Map