Assay Details
Target Gene Details
Entrez Gene ID: | 4701 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A7 |
Gene Aliases: |
B14.5a, CI-B14.5a |
Location: |
Chr.19:8308603-8321413 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA7 | NM_005001.4 | NP_004992.2 | ||
| NR_135539.1 | ||||
| AF050637.1 | AAD05427.1 | |||
| AF054178.1 | AAC99399.1 | |||
| BC003102.1 | AAH03102.1 | |||
| BC107892.1 | AAI07893.1 | |||
| CD172691.1 | ||||
| CR456915.1 | CAG33196.1 | |||
| CR542266.1 | CAG47062.1 | |||
| CR975510.1 | ||||
| DA189271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2401 | Chr.19:8258600 - 8320660 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map