Assay Details
Target Gene Details
Entrez Gene ID: | 10591 |
Gene Name: | 2'-deoxynucleoside 5'-phosphate N-hydrolase 1 |
Gene Aliases: |
C6orf108, RCL, dJ330M21.3 |
Location: |
Chr.6:43225629-43229473 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNPH1 | NM_006443.2 | NP_006434.1 | ||
| NM_199184.1 | 3 | 434 | NP_954653.1 | |
| AF040105.1 | AAB96766.1 | |||
| BC011683.2 | AAH11683.1 | |||
| BM747628.1 | ||||
| EU585603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602997 | Chr.6:43221202 - 43291349 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map