Assay Details
Target Gene Details
Entrez Gene ID: | 340156 |
Gene Name: | myosin light chain kinase family member 4 |
Gene Aliases: |
SgK085 |
Location: |
Chr.6:2663629-2770330 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK4 | NM_001012418.3 | NP_001012418.2 | ||
| XM_005249078.3 | XP_005249135.2 | |||
| XM_005249079.2 | XP_005249136.1 | |||
| XM_006715082.3 | XP_006715145.1 | |||
| XM_011514567.2 | XP_011512869.1 | |||
| XM_017010819.1 | XP_016866308.1 | |||
| BC132831.1 | ||||
| BC132833.1 | ||||
| BX647582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607900 | Chr.6:2093592 - 2775672 on Build GRCh38 | Gain |
|
| esv3607912 | Chr.6:2623291 - 2776818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map