Assay Details
Target Gene Details
Entrez Gene ID: | 4825 |
Gene Name: | NK6 homeobox 1 |
Gene Aliases: |
NKX6.1, NKX6A |
Location: |
Chr.4:84493283-84498234 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX6-1 | NM_006168.2 | 3 | 1046 | NP_006159.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829998 | Chr.4:84378540 - 84552297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map