Assay Details
Target Gene Details
Entrez Gene ID: | 5205 |
Gene Name: | ATPase phospholipid transporting 8B1 |
Gene Aliases: |
ATPIC, BRIC, FIC1, ICP1, PFIC, PFIC1 |
Location: |
Chr.18:57646426-57803828 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP8B1 | NM_005603.4 | NP_005594.1 | ||
| XM_006722481.3 | XP_006722544.1 | |||
| XM_011526022.2 | XP_011524324.1 | |||
| XM_011526023.2 | XP_011524325.1 | |||
| AF038007.1 | AAC63461.1 | |||
| BC003534.1 | AAH03534.1 |
Target Gene Details
Entrez Gene ID: | 100505549 |
Gene Name: | uncharacterized LOC100505549 |
Gene Aliases: |
- |
Location: |
Chr.18:57630302-57669276 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100505549 | NM_001242804.1 | NP_001229733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833663 | Chr.18:57549030 - 57718589 on Build GRCh38 | Gain |
|
| nsv2319 | Chr.18:57638082 - 57658017 on Build GRCh38 | Insertion |
|
| nsv458086 | Chr.18:57517516 - 57756707 on Build GRCh38 | Gain |
|
| esv2751777 | Chr.18:57515803 - 57805084 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map