Assay Details
Target Gene Details
Entrez Gene ID: | 339168 |
Gene Name: | transmembrane protein 95 |
Gene Aliases: |
UNQ9390 |
Location: |
Chr.17:7354968-7357219 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM95 | NM_001320435.1 | 1 | 131 | NP_001307364.1 |
| NM_001320436.1 | 1 | 131 | NP_001307365.1 | |
| NM_198154.2 | 1 | 131 | NP_937797.1 | |
| XM_017024565.1 | 1 | 153 | XP_016880054.1 | |
| XM_017024566.1 | 1 | 153 | XP_016880055.1 | |
| XM_017024567.1 | 1 | 153 | XP_016880056.1 | |
| XM_017024568.1 | 1 | 153 | XP_016880057.1 | |
| XM_017024569.1 | 1 | 153 | XP_016880058.1 | |
| XM_017024570.1 | XP_016880059.1 | |||
| XM_017024571.1 | 1 | 299 | XP_016880060.1 | |
| AY358218.1 | 1 | 89 | AAQ88585.1 | |
| BC040900.1 | 1 | 131 | AAH40900.1 | |
| BC107110.2 | 1 | 89 | AAI07111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|

Genomic Map