Assay Details
Target Gene Details
Entrez Gene ID: | 2305 |
Gene Name: | forkhead box M1 |
Gene Aliases: |
FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT |
Location: |
Chr.12:2857681-2877155 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXM1 | NM_001243088.1 | 8 | 1828 | NP_001230017.1 |
| NM_001243089.1 | 8 | 1825 | NP_001230018.1 | |
| NM_021953.3 | 9 | 1873 | NP_068772.2 | |
| NM_202002.2 | 10 | 1987 | NP_973731.1 | |
| NM_202003.2 | 8 | 1828 | NP_973732.1 | |
| XM_005253676.3 | 9 | 1876 | XP_005253733.1 | |
| XM_011520930.2 | 9 | 1873 | XP_011519232.1 | |
| XM_011520931.2 | 8 | 1831 | XP_011519233.1 | |
| XM_011520934.2 | 10 | 1934 | XP_011519236.1 | |
| AK291206.1 | 8 | 1728 | ||
| AK313845.1 | 8 | 1674 | ||
| AY542306.1 | 8 | 1641 | AAT08175.1 | |
| BC006192.2 | 8 | 1713 | AAH06192.1 | |
| BC006529.2 | 8 | 1713 | AAH06529.1 | |
| BC012863.1 | 8 | 1609 | AAH12863.1 | |
| BT006986.1 | 8 | 1590 | AAP35632.1 | |
| U74612.1 | 10 | 1818 | AAC51128.1 | |
| U74613.1 | 8 | 1652 | AAC51129.1 | |
| U83113.1 | 9 | 1659 | AAC63595.1 |
Target Gene Details
Entrez Gene ID: | 100507424 |
Gene Name: | uncharacterized LOC100507424 |
Gene Aliases: |
- |
Location: |
Chr.12:2836816-2859795 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507424 | NR_038933.1 | |||
| BU947711.1 | ||||
| BU947982.1 | ||||
| U90917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain |
|
| dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss |
|
| nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map