Assay Details
Target Gene Details
Entrez Gene ID: | 79959 |
Gene Name: | centrosomal protein 76 |
Gene Aliases: |
C18orf9, HsT1705 |
Location: |
Chr.18:12661733-12702777 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP76 | NM_001271989.1 | NP_001258918.1 | ||
| NM_024899.3 | NP_079175.2 | |||
| NR_073537.1 | ||||
| XM_005258149.4 | XP_005258206.1 | |||
| XM_017025981.1 | XP_016881470.1 | |||
| XM_017025982.1 | XP_016881471.1 | |||
| XM_017025983.1 | XP_016881472.1 | |||
| XM_017025984.1 | XP_016881473.1 | |||
| XM_017025985.1 | XP_016881474.1 | |||
| XM_017025986.1 | XP_016881475.1 | |||
| XM_017025987.1 | XP_016881476.1 | |||
| AK022604.1 | BAB14123.1 | |||
| AK301964.1 | ||||
| AK303117.1 | ||||
| AL833727.1 | CAH56252.1 | |||
| BC026307.1 | AAH26307.1 |
Target Gene Details
Entrez Gene ID: | 56984 |
Gene Name: | proteasome assembly chaperone 2 |
Gene Aliases: |
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1 |
Location: |
Chr.18:12658739-12725740 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMG2 | NM_147163.1 | NP_671692.1 | ||
| AF068295.1 | AAF65184.1 | |||
| AF161378.1 | AAF28938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| nsv523743 | Chr.18:12474506 - 12742810 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map