Assay Details
Target Gene Details
Entrez Gene ID: | 570 |
Gene Name: | bile acid-CoA:amino acid N-acyltransferase |
Gene Aliases: |
BACAT, BAT |
Location: |
Chr.9:101360417-101385005 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAAT | NM_001127610.1 | 4 | 2916 | NP_001121082.1 |
| NM_001701.3 | 4 | 3017 | NP_001692.1 | |
| CB154234.1 | 1 | 527 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037157 | Chr.9:101283479 - 101386951 on Build GRCh38 | Loss |
|
| nsv6641 | Chr.9:101343597 - 101365101 on Build GRCh38 | Insertion |
|
| nsv509314 | Chr.9:101354608 - 101368514 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map