Assay Details
Target Gene Details
Entrez Gene ID: | 94005 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class S |
Gene Aliases: |
- |
Location: |
Chr.17:28553388-28571869 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGS | NM_033198.3 | NP_149975.1 | ||
| AB057723.1 | BAB60853.1 | |||
| AK055512.1 | BAB70938.1 | |||
| AK075428.1 | BAC11615.1 | |||
| AK297046.1 | ||||
| AK314583.1 | ||||
| AY359112.1 | AAQ89470.1 | |||
| BC007301.1 | ||||
| BC069228.1 | AAH69228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
| nsv1064958 | Chr.17:28490519 - 28565886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map