Assay Details
Target Gene Details
Entrez Gene ID: | 2064 |
Gene Name: | erb-b2 receptor tyrosine kinase 2 |
Gene Aliases: |
CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1 |
Location: |
Chr.17:39688084-39728662 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERBB2 | NM_001005862.2 | NP_001005862.1 | ||
| NM_001289936.1 | NP_001276865.1 | |||
| NM_001289937.1 | NP_001276866.1 | |||
| NM_004448.3 | NP_004439.2 | |||
| NR_110535.1 | ||||
| AK295195.1 | ||||
| AK300321.1 | ||||
| AK313683.1 | ||||
| BC167147.1 | ||||
| KJ534831.1 | ||||
| KJ534832.1 | ||||
| KJ534964.1 | ||||
| M11730.1 | AAA75493.1 | |||
| X03363.1 | CAA27060.1 |
Target Gene Details
Entrez Gene ID: | 100616132 |
Gene Name: | microRNA 4728 |
Gene Aliases: |
- |
Location: |
Chr.17:39726495-39726561 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4728 | NR_039881.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056234 | Chr.17:39706352 - 39753735 on Build GRCh38 | Gain |
|
| nsv2048 | Chr.17:39718682 - 39742588 on Build GRCh38 | Insertion |
|
| dgv40n68 | Chr.17:39536627 - 39726611 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map