Assay Details
Target Gene Details
Entrez Gene ID: | 387849 |
Gene Name: | RAB15 effector protein |
Gene Aliases: |
RAB15EP |
Location: |
Chr.12:27696495-27697633 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REP15 | NM_001029874.1 | 1 | 931 | NP_001025045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832358 | Chr.12:27519016 - 27705439 on Build GRCh38 | Gain |
|
| esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
|
| nsv557823 | Chr.12:27695850 - 27762012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map