Assay Details
Target Gene Details
Entrez Gene ID: | 10498 |
Gene Name: | coactivator associated arginine methyltransferase 1 |
Gene Aliases: |
PRMT4 |
Location: |
Chr.19:10871577-10923078 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARM1 | NM_199141.1 | 16 | 2765 | NP_954592.1 |
| XM_005259708.4 | 15 | 2662 | XP_005259765.1 | |
| XM_011527638.2 | 15 | 2521 | XP_011525940.1 | |
| AF055027.1 | 11 | 1940 | ||
| AL833242.1 | 16 | 2558 |
Target Gene Details
Entrez Gene ID: | 78992 |
Gene Name: | Yip1 domain family member 2 |
Gene Aliases: |
FinGER2 |
Location: |
Chr.19:10922185-10929012 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF2 | XM_011528270.2 | 10 | 1780 | XP_011526572.1 |
| XM_011528271.2 | 9 | 1624 | XP_011526573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833749 | Chr.19:10881255 - 11027578 on Build GRCh38 | Loss |
|
| nsv833748 | Chr.19:10757002 - 10925368 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map