Assay Details
Target Gene Details
Entrez Gene ID: | 59340 |
Gene Name: | histamine receptor H4 |
Gene Aliases: |
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R |
Location: |
Chr.18:24460568-24487427 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRH4 | NM_001143828.1 | 2 | 739 | NP_001137300.1 |
| NM_001160166.1 | 2 | 839 | NP_001153638.1 | |
| NM_021624.3 | 3 | 1003 | NP_067637.2 | |
| XM_011526133.1 | XP_011524435.1 | |||
| AB044934.1 | 3 | 957 | BAB13698.1 | |
| AB045370.1 | 3 | 966 | BAB20091.1 | |
| AF307973.1 | 3 | 903 | AAG32052.1 | |
| AF312230.1 | 3 | 1003 | AAK12081.1 | |
| AF325356.1 | 3 | 903 | AAL01684.1 | |
| AF329449.1 | 3 | 903 | AAK43542.1 | |
| AJ298292.1 | 3 | 903 | CAC83493.1 | |
| AY008280.1 | 3 | 903 | AAL09297.1 | |
| AY136745.1 | 3 | 903 | AAN01271.1 | |
| BC069136.1 | 3 | 1003 | AAH69136.1 | |
| BC112348.1 | 3 | 963 | AAI12349.1 | |
| DQ835186.1 | 2 | 639 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2716869 | Chr.18:24474907 - 24479209 on Build GRCh38 | Deletion |
|
| nsv521369 | Chr.18:24468583 - 24479130 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map