Assay Details
Target Gene Details
Entrez Gene ID: | 90952 |
Gene Name: | endothelial cell adhesion molecule |
Gene Aliases: |
W117m |
Location: |
Chr.11:124753123-124762327 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESAM | NM_138961.2 | 7 | 1110 | NP_620411.2 |
| XM_005271720.4 | 4 | 531 | XP_005271777.1 | |
| XM_011543060.2 | 6 | 873 | XP_011541362.1 | |
| AF277292.1 | AAF97236.1 | |||
| AF361746.1 | 7 | 1075 | AAK51065.1 | |
| AK075396.1 | 7 | 1073 | BAC11594.1 | |
| AK091023.1 | 7 | 2004 | ||
| AK092429.1 | 5 | 1482 | ||
| AK301160.1 | ||||
| AK314710.1 | 7 | 1084 | ||
| AY189281.1 | 4 | 1667 | AAO86725.1 | |
| AY358382.1 | 7 | 1062 | AAQ88748.1 | |
| BC016868.1 | 7 | 944 | AAH16868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556487 | Chr.11:124730862 - 124765248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map