Assay Details
Target Gene Details
Entrez Gene ID: | 4015 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
- |
Location: |
Chr.5:122063195-122078501 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOX | NM_001178102.2 | 6 | 1189 | NP_001171573.1 |
| NM_001317073.1 | 6 | 941 | NP_001304002.1 | |
| NM_002317.6 | 7 | 2114 | NP_002308.2 | |
| AF039291.1 | 7 | 1847 | AAD02130.1 | |
| AK297709.1 | 8 | 1738 | ||
| AK298781.1 | ||||
| AK307559.1 | 6 | 946 | ||
| AK307732.1 | 6 | 941 | ||
| AK315873.1 | 6 | 924 | ||
| AK316309.1 | 6 | 1077 | ||
| BC089436.1 | AAH89436.1 |
Target Gene Details
Entrez Gene ID: | 153443 |
Gene Name: | serum response factor binding protein 1 |
Gene Aliases: |
BUD22, P49, Rlb1, STRAP, p49/STRAP |
Location: |
Chr.5:121961917-122075624 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRFBP1 | XM_017009111.1 | XP_016864600.1 |
More Information
Set Membership: |
|

Genomic Map