Assay Details
Target Gene Details
Entrez Gene ID: | 9946 |
Gene Name: | crystallin zeta like 1 |
Gene Aliases: |
4P11, QOH-1 |
Location: |
Chr.21:33589341-33641854 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYZL1 | NM_145858.2 | NP_665857.2 | ||
| BC033023.2 | AAH33023.1 | |||
| CA426424.1 |
More Information
Set Membership: |
|

Genomic Map