Assay Details
Target Gene Details
Entrez Gene ID: | 2091 |
Gene Name: | fibrillarin |
Gene Aliases: |
FIB, FLRN, RNU3IP1 |
Location: |
Chr.19:39834453-39846414 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBL | NM_001436.3 | NP_001427.2 | ||
| XM_005258651.2 | XP_005258708.1 | |||
| XM_011526623.2 | XP_011524925.1 | |||
| AB451384.1 | ||||
| AI085380.1 | ||||
| AK296950.1 | ||||
| BC015218.2 | AAH15218.2 | |||
| BC019260.1 | AAH19260.1 | |||
| BC019609.1 | AAH19609.1 | |||
| BT006830.1 | AAP35476.1 | |||
| BT020144.1 | AAV38946.1 | |||
| CR457069.1 | CAG33350.1 | |||
| M59849.1 | AAA52453.1 | |||
| X56597.1 | CAA39935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758760 | Chr.19:39737264 - 40015223 on Build GRCh38 | Gain |
|
| nsv9725 | Chr.19:39824879 - 39906616 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map