Assay Details
Target Gene Details
Entrez Gene ID: | 441024 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like |
Gene Aliases: |
- |
Location: |
Chr.4:74123280-74303099 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTHFD2L | NM_001144978.1 | 2 | 228 | NP_001138450.1 |
| XM_005265690.2 | 3 | 271 | XP_005265747.1 | |
| XM_017008218.1 | 2 | 608 | XP_016863707.1 | |
| XM_017008219.1 | 2 | 608 | XP_016863708.1 | |
| XM_017008220.1 | 2 | 608 | XP_016863709.1 | |
| XM_017008221.1 | 2 | 608 | XP_016863710.1 | |
| XM_017008222.1 | 2 | 608 | XP_016863711.1 | |
| XM_017008223.1 | 2 | 608 | XP_016863712.1 | |
| XM_017008224.1 | 2 | 1833 | XP_016863713.1 | |
| XM_017008225.1 | 5 | 389 | XP_016863714.1 | |
| XM_017008226.1 | 5 | 264 | XP_016863715.1 | |
| XM_017008227.1 | 3 | 271 | XP_016863716.1 | |
| XM_017008228.1 | 3 | 271 | XP_016863717.1 | |
| XM_017008229.1 | 3 | 271 | XP_016863718.1 | |
| XM_017008230.1 | 3 | 271 | XP_016863719.1 | |
| XM_017008231.1 | 4 | 314 | XP_016863720.1 | |
| XM_017008232.1 | 3 | 280 | XP_016863721.1 | |
| XM_017008234.1 | 3 | 271 | XP_016863723.1 | |
| XM_017008235.1 | 3 | 233 | XP_016863724.1 | |
| AI219460.1 | 2 | 167 | ||
| BC032771.1 | 3 | 187 | AAH32771.1 | |
| BC037529.2 | 3 | 266 | AAH37529.2 | |
| BG708427.1 | 3 | 271 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569519 | Chr.4:74142954 - 74255926 on Build GRCh38 | Loss |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
| dgv9134n54 | Chr.4:74147724 - 74240768 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map