Assay Details
Target Gene Details
Entrez Gene ID: | 64858 |
Gene Name: | DNA cross-link repair 1B |
Gene Aliases: |
APOLLO, SNM1B, SNMIB |
Location: |
Chr.1:113904619-113914086 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCLRE1B | NM_001319946.1 | 1 | 162 | NP_001306875.1 |
| NM_001319947.1 | NP_001306876.1 | |||
| NM_022836.3 | 1 | 162 | NP_073747.1 | |
| AF306691.1 | 1 | 46 | ||
| AF306693.1 | 1 | 97 | ||
| AK024060.1 | 1 | 80 | BAB14807.1 | |
| BC029687.1 | 1 | 39 | AAH29687.1 | |
| CR936679.1 | 1 | 87 | ||
| DA414122.1 | ||||
| DR005221.1 | 1 | 162 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587251 | Chr.1:113871703 - 113936287 on Build GRCh38 | Loss |
|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map