Assay Details
Target Gene Details
Entrez Gene ID: | 5452 |
Gene Name: | POU class 2 homeobox 2 |
Gene Aliases: |
OCT2, OTF2, Oct-2 |
Location: |
Chr.19:42086110-42197931 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU2F2 | NM_001207025.2 | 14 | 2495 | NP_001193954.1 |
| NM_001247994.1 | 14 | 2311 | NP_001234923.1 | |
| NM_002698.4 | 14 | 2447 | NP_002689.1 | |
| XM_005259010.3 | 15 | 2538 | XP_005259067.1 | |
| XM_011527040.2 | 15 | 2652 | XP_011525342.1 | |
| XM_011527041.2 | 14 | 2578 | XP_011525343.1 | |
| XM_011527042.2 | 14 | 2530 | XP_011525344.1 | |
| XM_011527043.2 | 15 | 2739 | XP_011525345.2 | |
| XM_017026884.1 | 16 | 2950 | XP_016882373.1 | |
| XM_017026885.1 | 16 | 2953 | XP_016882374.1 | |
| XM_017026886.1 | 17 | 4452 | XP_016882375.1 | |
| XM_017026887.1 | 17 | 2922 | XP_016882376.1 | |
| XM_017026888.1 | 17 | 3402 | XP_016882377.1 | |
| XM_017026889.1 | 16 | 2944 | XP_016882378.1 | |
| XM_017026890.1 | 16 | 3025 | XP_016882379.1 | |
| XM_017026891.1 | 16 | 2902 | XP_016882380.1 | |
| XM_017026892.1 | 16 | 2884 | XP_016882381.1 | |
| XM_017026893.1 | 15 | 2604 | XP_016882382.1 | |
| XM_017026894.1 | 15 | 2586 | XP_016882383.1 | |
| XM_017026895.1 | 15 | 2584 | XP_016882384.1 | |
| XM_017026896.1 | 15 | 2876 | XP_016882385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map