Assay Details
Target Gene Details
Entrez Gene ID: | 53827 |
Gene Name: | FXYD domain containing ion transport regulator 5 |
Gene Aliases: |
DYSAD, HSPC113, IWU1, KCT1, OIT2, PRO6241, RIC |
Location: |
Chr.19:35154722-35169885 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FXYD5 | NM_001164605.1 | NP_001158077.1 | ||
| NM_001320912.1 | 8 | 560 | NP_001307841.1 | |
| NM_001320913.1 | NP_001307842.1 | |||
| NM_014164.5 | NP_054883.3 | |||
| NM_144779.2 | NP_659003.1 | |||
| NR_028406.1 | ||||
| AA044211.1 | ||||
| AB072911.1 | BAB83766.1 | |||
| AF161462.1 | AAF29077.1 | |||
| AF177940.1 | AAG09301.1 | |||
| AI424071.1 | ||||
| AK304012.1 | 8 | 552 | ||
| AY157582.1 | AAO13166.1 | |||
| AY358991.1 | AAQ89350.1 | |||
| BC009642.2 | AAH09642.1 | |||
| BG025158.1 | ||||
| BM550889.1 | 8 | 495 | ||
| BQ183683.1 | ||||
| BQ440098.1 | ||||
| BQ941279.1 | ||||
| BT007343.1 | AAP36007.1 | |||
| BU164524.1 | ||||
| BU178888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758757 | Chr.19:35162739 - 35182430 on Build GRCh38 | Loss |
|
| nsv2470 | Chr.19:35164378 - 35211662 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map