Assay Details
Target Gene Details
Entrez Gene ID: | 4745 |
Gene Name: | neural EGFL like 1 |
Gene Aliases: |
IDH3GL, NRP1 |
Location: |
Chr.11:20669551-21575686 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NELL1 | NM_001288713.1 | NP_001275642.1 | ||
| NM_001288714.1 | NP_001275643.1 | |||
| NM_006157.4 | NP_006148.2 | |||
| NM_201551.2 | NP_963845.1 | |||
| AK127805.1 | ||||
| AK299558.1 | ||||
| AK303736.1 | ||||
| AK313445.1 | ||||
| BC069674.1 | AAH69674.1 | |||
| BC096100.1 | AAH96100.1 | |||
| BC096101.1 | AAH96101.1 | |||
| BC096102.3 | AAH96102.1 | |||
| BC096103.3 | AAH96103.1 | |||
| BX476569.1 | ||||
| D83017.1 | BAA11680.1 | |||
| DA165320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7703 | Chr.11:20632781 - 20680091 on Build GRCh38 | Deletion |
|
| nsv553678 | Chr.11:20097752 - 20718575 on Build GRCh38 | Gain |
|
| nsv553680 | Chr.11:20668471 - 20738448 on Build GRCh38 | Loss |
|
| esv3579418 | Chr.11:20665173 - 20673473 on Build GRCh38 | Loss |
|
| nsv519547 | Chr.11:20667940 - 20680488 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map