Assay Details
Target Gene Details
Entrez Gene ID: | 3321 |
Gene Name: | immunoglobulin superfamily member 3 |
Gene Aliases: |
EWI-3, LCDD, V8 |
Location: |
Chr.1:116574398-116667755 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF3 | NM_001007237.2 | 10 | 3739 | NP_001007238.1 |
| NM_001542.3 | 10 | 3661 | NP_001533.2 | |
| XM_005270793.2 | 11 | 3771 | XP_005270850.1 | |
| XM_005270794.4 | 9 | 3009 | XP_005270851.1 | |
| XM_006710593.3 | 10 | 3072 | XP_006710656.1 | |
| XM_011541315.1 | 11 | 3823 | XP_011539617.1 | |
| XM_011541316.2 | 5 | 2552 | XP_011539618.1 | |
| AB007935.2 | 9 | 3071 | ||
| AF031174.1 | 10 | 3136 | AAC72013.1 | |
| BC110651.1 | 10 | 3739 | AAI10652.1 | |
| BC130279.1 | 9 | 3409 | ||
| BC144132.1 | 9 | 3409 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482103 | Chr.1:116574409 - 116667692 on Build GRCh38 | Gain |
|
| esv3578131 | Chr.1:116577298 - 116581814 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map