Assay Details
Target Gene Details
Entrez Gene ID: | 9480 |
Gene Name: | one cut homeobox 2 |
Gene Aliases: |
OC-2, OC2 |
Location: |
Chr.18:57435420-57491298 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ONECUT2 | NM_004852.2 | 1 | 440 | NP_004843.2 |
| XM_017026096.1 | 1 | 705 | XP_016881585.1 | |
| Y18198.1 | 1 | 440 | CAB38253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1614n106 | Chr.18:57435368 - 57436168 on Build GRCh38 | Deletion |
|
| nsv828250 | Chr.18:57424010 - 57442352 on Build GRCh38 | Gain |
|
| nsv953552 | Chr.18:57429769 - 57447868 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map