Assay Details
Target Gene Details
Entrez Gene ID: | 126637 |
Gene Name: | trichohyalin like 1 |
Gene Aliases: |
S100A17, THHL1 |
Location: |
Chr.1:152084144-152089064 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCHHL1 | NM_001008536.1 | 3 | 570 | NP_001008536.1 |
| AY456639.1 | 3 | 570 | AAS13485.1 | |
| BC137493.1 | 3 | 560 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
| nsv547853 | Chr.1:152036195 - 152264284 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77167778] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map