Assay Details
Target Gene Details
Entrez Gene ID: | 23432 |
Gene Name: | G protein-coupled receptor 161 |
Gene Aliases: |
RE2 |
Location: |
Chr.1:168079542-168137667 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32660 | Chr.1:167825845 - 168220986 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map