Assay Details
Target Gene Details
Entrez Gene ID: | 9567 |
Gene Name: | GTP binding protein 1 |
Gene Aliases: |
GP-1, GP1, HSPC018 |
Location: |
Chr.22:38704561-38738265 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP1 | XM_011530537.2 | XP_011528839.1 | ||
| XM_017029099.1 | 14 | 3506 | XP_016884588.1 | |
| XM_017029100.1 | 14 | 3476 | XP_016884589.1 | |
| XM_017029101.1 | 15 | 2093 | XP_016884590.1 | |
| XM_017029102.1 | 13 | 1909 | XP_016884591.1 | |
| XM_017029103.1 | 13 | 2007 | XP_016884592.1 |
Target Gene Details
Entrez Gene ID: | 25777 |
Gene Name: | Sad1 and UNC84 domain containing 2 |
Gene Aliases: |
UNC84B |
Location: |
Chr.22:38734714-38756019 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUN2 | NM_001199579.1 | 18 | 3546 | NP_001186508.1 |
| NM_001199580.1 | 19 | 3482 | NP_001186509.1 | |
| NM_015374.2 | 18 | 3483 | NP_056189.1 | |
| XM_011530104.2 | 17 | 3340 | XP_011528406.1 | |
| XM_011530105.2 | 18 | 3207 | XP_011528407.1 | |
| AB014568.1 | 18 | 3265 | ||
| AF038178.1 | 1 | 1110 | ||
| BC030684.2 | 6 | 1688 | AAH30684.2 | |
| BC069253.1 | 10 | 2364 | AAH69253.1 | |
| BC094797.1 | 18 | 3503 | AAH94797.1 | |
| BC111549.1 | 17 | 2905 | AAI11550.1 | |
| BC111717.1 | 17 | 3212 | AAI11718.1 | |
| BE675617.1 | ||||
| BX537962.1 | 19 | 3474 | CAD97926.1 | |
| CD365855.1 | 1 | 519 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv991516 | Chr.22:38729936 - 38740885 on Build GRCh38 | Deletion |
|
| dgv769n67 | Chr.22:38729402 - 38889280 on Build GRCh38 | Gain |
|
| nsv1055817 | Chr.22:38729069 - 38767021 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map